Arbutin 98%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxyphenyl beta-D-Glucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(4-hydroxyphenoxy)tetrahydro-2H-pyran-3,4,5-triol, Hydroquinone-beta-D-glucoside

A309050-10G A309050-1G A309050-25G A309050-500G A309050-5G
76723-798EA 9 USD
76723-798 76723-800 76723-802 76723-804 76723-806
Arbutin 98%
Arbutin

Caution: for Research Use Only

Formula: C₁₂H₁₆O₇
MW: 272.25 g/mol
MDL Number: MFCD00016915
CAS Number: 497-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR