Mizoribine 98%

Supplier: Ambeed
Warning

Synonyms: 1-(beta-D-Ribofuranosyl)-5-hydroxyimidazole-4-carboxamide, 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-hydroxy-1H-imidazole-4-carboxamide

A306960-100MG A306960-10MG A306960-1G A306960-1MG A306960-50MG A306960-5MG
76723-754EA 53 USD
76723-754 76723-756 76723-758 76723-760 76723-762 76723-764
Mizoribine 98%
Mizoribine

Caution: for Research Use Only

Formula: C₉H₁₃N₃O₆
MW: 259.22 g/mol
MDL Number: MFCD00057221
CAS Number: 50924-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR