2-Isopropyl-N,2,3-trimethylbutyramide 97%
Supplier: Ambeed
|
Warning
|
A288487-100G
A288487-10G
A288487-25G
A288487-500G
A288487-5G
76723-436EA
66
USD
76723-436
76723-438
76723-440
76723-442
76723-444
76723-446
2-Isopropyl-N,2,3-trimethylbutyramide 97%
2-Isopropyl-N,2,3-trimethylbutyramide
Caution: for Research Use Only
Formula:
C₁₀H₂₁NO |
MDL Number:
MFCD00130070 CAS Number: 51115-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...