Octadecylphosphonic acid 98%

Supplier: Ambeed
Warning

Synonyms: n-Octadecylphosphonic acid, ODPA

A281298-100G A281298-25G A281298-5G
76723-284EA 41 USD
76723-284 76723-286 76723-288
Octadecylphosphonic acid 98%
Octadecylphosphonic acid

Caution: for Research Use Only

Formula: C₁₈H₃₉O₃P
MDL Number: MFCD00015838
CAS Number: 4724-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR