Naphthacene 98%

Supplier: Ambeed
Danger

Synonyms: 2,3-Benzanthracene, Benz[b]anthracene, Tetracene

A279269-100MG A279269-1G A279269-250MG A279269-5G
76723-234EA 46 USD
76723-234 76723-236 76723-238 77568-556
Naphthacene 98%
Naphthacene

Caution: For research use only.

Formula: C₁₈H₁₂
MW: 228.29 g/mol
Boiling Pt: 437
Melting Pt: 356
MDL Number: MFCD00003702
CAS Number: 92-24-0
UN: 3077
ADR: 9 Ⅲ
Merck Index: 12,06456

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR