Picolinamide 98%

Supplier: Ambeed
Warning

Synonyms: 2-Pyridinecarboxamide, Pyridine-2-carboxamide

A226558-100G A226558-10G A226558-1G A226558-250MG A226558-25G A226558-5G
76722-122EA 52 USD
76722-122 76722-124 76722-126 76722-128 76722-130 76722-132
Picolinamide 98%
Picolinamide

Caution: for Research Use Only

Formula: C₆H₆N₂O
MW: 122.13 g/mol
MDL Number: MFCD00023483
CAS Number: 1452-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR