(±)-1,2-Propanediol 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane
A205936-1000G
A205936-100G
A205936-25G
A205936-500G
76721-554EA
19
USD
76721-554
76721-556
76721-558
76721-560
(±)-1,2-Propanediol 99%
Propylene glycol
Caution: for Research Use Only
Formula:
CH₃CH(OH)CH₂OH MW: 76.1 g/mol Boiling Pt: 187.6 Melting Pt: -59 Flash Pt: 99 |
MDL Number:
MFCD00064272 CAS Number: 57-55-6 Merck Index: 13,07947 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...