Potassium trifluoromethanesulfonate 95%
Supplier: Ambeed
|
Warning
|
A180593-100G
A180593-1G
A180593-25G
A180593-5G
76720-854EA
72
USD
76720-854
76720-856
76720-858
76720-860
Potassium trifluoromethanesulfonate 95%
Potassium trifluoromethanesulphonate
Caution: for Research Use Only
Formula:
CF₃KO₃S MW: 188.17 g/mol |
MDL Number:
MFCD00042370 CAS Number: 2926-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...