Bergapten 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methoxypsoralen, 4-Methoxyfuro[3,2-g]benzopyran-7-one, 4-Methoxy-7H-furo[3,2-g]chromen-7-one

A141454-1G A141454-1MG A141454-250MG A141454-5G A141454-10G A141454-25G
76719-384EA 53.05 USD
76719-384 76719-386 76719-388 76719-390 77600-056 77667-054
Bergapten 98%
Bergapten

Caution: for Research Use Only

Formula: C₁₂H₈O₄
MW: 216.19 g/mol
MDL Number: MFCD00010272
CAS Number: 484-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR