Posaconazole 98%
Supplier: Ambeed
|
Warning
|
A133379-10G
A133379-10MG
A133379-1G
A133379-50MG
A133379-5G
A133379-25G
A133379-100G
A133379-100MG
A133379-250MG
A133379-5MG
76719-022EA
219
USD
76719-022
76719-024
76719-026
76719-028
76719-030
77592-534
77599-910
77599-912
77599-914
77666-914
Posaconazole 98%
Posaconazole
Caution: for Research Use Only
Formula:
C₃₇H₄₂F₂N₈O₄ |
MDL Number:
MFCD00941162 CAS Number: 171228-49-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...