4'-Methoxybutyrophenone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(4-Methoxyphenyl)butan-1-one
A122660-100G
A122660-1G
A122660-25G
A122660-5G
76718-620EA
361
USD
76718-620
76718-622
76718-624
77666-758
4'-Methoxybutyrophenone 98%
4'-Methoxybutyrophenone
Caution: for Research Use Only
Formula:
C₁₁H₁₄O₂ |
MDL Number:
MFCD00027138 CAS Number: 4160-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...