Pyridinium p-toluenesulfonate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pyridin-1-ium 4-methylbenzenesulfonate, Pyridin-1-ium 4-methylbenzenesulphonate
A122417-1000G
A122417-100G
A122417-10G
A122417-1G
A122417-25G
A122417-500G
A122417-5G
76718-600EA
131
USD
76718-600
76718-602
76718-604
76718-606
76718-608
76718-610
76718-612
Pyridinium p-toluenesulfonate 98%
Pyridinium p-toluenesulphonate
Caution: for Research Use Only
Formula:
C₁₂H₁₃NO₃S MW: 251.31 g/mol |
MDL Number:
MFCD00013108 CAS Number: 24057-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...