1-Monodecanoin 95%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydroxypropyl decanoate

A111808-1G A111808-250MG A111808-5G
76718-110EA 56 USD
76718-110 76718-112 77878-842
1-Monodecanoin 95%
1-Monodecanoin

Caution: for Research Use Only

Formula: C₁₃H₂₆O₄
MDL Number: MFCD00056656
CAS Number: 2277-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR