Sebacic acid 95%

Supplier: Ambeed
Warning

A109865-100G A109865-10G A109865-25G A109865-500G A109865-1000G
76718-040EA 8 USD
76718-040 76718-042 76718-044 76718-046 77549-122
Sebacic acid 95%
Sebacic acid

Caution: for Research Use Only

Formula: C₁₀H₁₈O₄
MDL Number: MFCD00004440
CAS Number: 111-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR