Benzyl benzoate 99%

Supplier: Ambeed
Danger

Synonyms: Benzoic acid benzyl ester

A954638-500G A954638-25G A954638-100G
76698-838EA 10 USD
76698-838 77569-684 77627-782
Benzyl benzoate 99%
Benzyl benzoate

Caution: For research use only.

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 323
Melting Pt: 21
Flash Pt: 158
MDL Number: MFCD00003075
CAS Number: 120-51-4
UN: 3082
ADR: 9 Ⅲ
Merck Index: 13,01129

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR