Coronene 98%

Supplier: Ambeed
Warning

Synonyms: Hexabenzobenzene

A921798-100MG A921798-1G A921798-250MG A921798-5G
76698-700EA 30.77 USD
76698-700 76698-702 76698-704 77573-842
Coronene 98%
Coronene

Caution: for Research Use Only

Formula: C₂₄H₁₂
MW: 300.36 g/mol
MDL Number: MFCD00004134
CAS Number: 191-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR