Aniracetam 97%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Methoxybenzoyl)pyrrolidin-2-one, 1-(4-Methoxybenzoyl)-2-pyrrolidone

A890956-100G A890956-100MG A890956-1G A890956-1MG A890956-250MG A890956-25G A890956-5G
76698-544EA 982.4 USD
76698-544 76698-546 76698-548 76698-550 76698-552 76698-554 76698-556
Aniracetam 97%
Aniracetam

Caution: for Research Use Only

Formula: C₁₂H₁₃NO₃
MW: 219.24 g/mol
MDL Number: MFCD00153767
CAS Number: 72432-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR