9,10-Anthracenediyl-bis(methylene)dimalonic acid 95%

Supplier: Ambeed
Danger

Synonyms: 2,2'-(Anthracene-9,10-diylbis(methylene))dimalonic acid

A888564-50MG A888564-100MG A888564-250MG A888564-1G
76698-542EA 49 USD
76698-542 77627-712 77627-714 77878-719
9,10-Anthracenediyl-bis(methylene)dimalonic acid 95%
9,10-Anthracenediyl-bis(methylene)dimalonic acid

Caution: for Research Use Only

Formula: C₂₂H₁₈O₈
MDL Number: MFCD03095464
CAS Number: 307554-62-7
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR