Conduritol B epoxide 97%

Supplier: Ambeed
Warning

Synonyms: rel-(1R,2R,3S,4S,5R,6S)-7-Oxabicyclo[4.1.0]heptane-2,3,4,5-tetraol

A815375-100MG A815375-10MG A815375-1MG A815375-25MG A815375-5MG A815375-250MG A815375-1G
76698-138EA 189 USD
76698-138 76698-140 76698-142 76698-144 76698-146 77603-062 77627-610
Conduritol B epoxide 97%
Conduritol B epoxide

Caution: for Research Use Only

Formula: C₆H₁₀O₅
MW: 162.141 g/mol
MDL Number: MFCD00077326
CAS Number: 6090-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR