Ethyl-N-acetylglycinate(Ac-Gly-OEt) 98%

Supplier: Ambeed
Warning

Synonyms: N-Acetylglycine ethyl ester, Ethyl 2-(acetylamino)acetate, Ac-Gly-OEt, Ethyl acetamidoacetate, Ethyl N-acetylglycinate

A677513-100G A677513-10G A677513-25G A677513-5G
76697-176EA 125.19 USD
76697-176 76697-178 76697-180 76697-182
Ethyl-N-acetylglycinate(Ac-Gly-OEt) 98%
Ethyl N-acetylglycinate (Ac-Gly-OEt)

Caution: for Research Use Only

Formula: C₆H₁₁NO₃
MW: 145.1 g/mol
MDL Number: MFCD00009173
CAS Number: 1906-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR