Coumarin 99+%

Supplier: Ambeed
Danger

Synonyms: 1-Benzopyran-2-one, 2H-Chromen-2-one, 1,2-Benzopyrone

A633268-1000G A633268-100G A633268-100MG A633268-10G A633268-25G A633268-500G
76696-822EA 49 USD
76696-822 76696-824 76696-826 76696-828 76696-830 76696-832
Coumarin 99+%
Coumarin

Caution: for Research Use Only

Formula: C₉H₆O₂
MDL Number: MFCD00006850
CAS Number: 91-64-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR