Copper(I) chloride 99%

Supplier: Ambeed
Danger

Synonyms: Cuprous chloride, Copper chloride

A631746-25G A631746-100G
76696-802EA 17 USD
76696-802 77627-264
Copper(I) chloride 99%
Copper (I) chloride

Caution: for Research Use Only

Formula: ClCu
MW: 99 g/mol
Boiling Pt: 1367
Melting Pt: 430
MDL Number: MFCD00010971
CAS Number: 7758-89-6
UN: 2802
ADR: 8 Ⅲ
Merck Index: 13,02690

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR