Phthalocyanine chloroaluminum ~85 % (dye content)

Supplier: Ambeed
Warning

Synonyms: Chloroaluminum Phthalocyanine

A580018-100G A580018-25G A580018-5G
76696-408EA 231.28 USD
76696-408 76696-410 76696-412
Phthalocyanine chloroaluminum ~85 % (dye content)
Phthalocyanine chloroaluminum

Caution: for Research Use Only

Formula: C₃₂H₁₆AlClN₈
MW: 574.97 g/mol
MDL Number: MFCD00049386
CAS Number: 14154-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR