Acetylacetonato(1,5-cyclooctadiene)rhodium(I) 98%

Supplier: Ambeed
Warning

Synonyms: (Acetylacetonato)(1,5-cyclooctadiene)rhodium(I)

A567452-1G A567452-250MG
76696-266EA 483 USD
76696-266 76696-268
Acetylacetonato(1,5-cyclooctadiene)rhodium(I) 98%
Acetylacetonato(1,5-cyclooctadiene)rhodium(I)

Caution: for Research Use Only

Formula: C₁₃H₂₀O₂Rh
MDL Number: MFCD00075046
CAS Number: 12245-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR