Sodium decanoate 98%

Supplier: Ambeed
Warning

A509712-100G A509712-1G A509712-25G A509712-500G A509712-5G
76695-746EA 137.92 USD
76695-746 76695-748 76695-750 76695-752 76695-754
Sodium decanoate 98%
Sodium decanoate

Caution: for Research Use Only

Formula: C₁₀H₁₉NaO₂
MW: 194.2464 g/mol
MDL Number: MFCD00066453
CAS Number: 1002-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR