Cyclohexanecarbonitrile 97%

Supplier: Ambeed
Danger

A497810-10G A497810-1G A497810-250MG A497810-25G A497810-5G
76695-682EA 39 USD
76695-682 76695-684 76695-686 76695-688 76695-690
Cyclohexanecarbonitrile 97%
Cyclohexanecarbonitrile

Caution: for Research Use Only

Formula: C₇H₁₁N
MW: 109.17 g/mol
MDL Number: MFCD00040444
CAS Number: 766-05-2
UN: 3276
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR