Butyramide 98%

Supplier: Ambeed
Warning

A491583-100G A491583-25G A491583-500G
76695-644EA 41 USD
76695-644 76695-646 76695-648
Butyramide 98%
Butyramide

Caution: for Research Use Only

Formula: C₄H₉NO
MDL Number: MFCD00041894
CAS Number: 541-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR