Benzophenone hydrazone 95%

Supplier: Ambeed
Danger

A490182-1000G A490182-100G A490182-25G A490182-500G
76695-620EA 65 USD
76695-620 76695-622 76695-624 76695-626 76695-628
Benzophenone hydrazone 95%
Benzophenone hydrazone

Caution: for Research Use Only

Formula: C₁₃H₁₂N₂
MDL Number: MFCD00007624
CAS Number: 5350-57-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR