2,2-Bis(4-glycidyloxyphenyl)propane 85%, Technical Grade
Supplier: Ambeed
|
Warning
|
Synonyms:
Bisphenol A Diglycidyl Ether, 2,2-Bis(4-hydroxyphenyl)propane Diglycidyl Ether, DER 332, BADGE, D.E.R. 332 epoxy resin
A422977-25G
A422977-100G
76694-968EA
13
USD
76694-968
77626-846
2,2-Bis(4-glycidyloxyphenyl)propane 85%, Technical Grade
2,2-Bis(4-glycidyloxyphenyl)propane
Caution: for Research Use Only
Formula:
C₂₁H₂₄O₄ MW: 340.42 g/mol |
MDL Number:
MFCD00080480 CAS Number: 1675-54-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...