(R)-(+)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol ≥97%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (R)-(+)-2,2'-Dihydroxy-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl, (R)-5,5',6,6',7,7',8,8'-Octahydro[1,1'-binaphthalene]-2,2'-diol

A412800-100G A412800-1G A412800-25G A412800-500G A412800-5G
76694-858EA 310 USD
76694-858 76694-860 76694-862 76694-864 76694-866 76694-868
(R)-(+)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol ≥97%, ee 99%
(R)-(+)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol

Caution: for Research Use Only

Formula: C₂₀H₂₂O₂
MDL Number: MFCD02093485
CAS Number: 65355-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR