Cesium sulfate 99%

Supplier: Ambeed
Warning

A388243-100G A388243-500G A388243-25G
76694-612EA 71 USD
76694-612 76694-614 77879-835
Cesium sulfate 99%
Cesium sulphate

Caution: for Research Use Only

Formula: Cs₂O₄S
MDL Number: MFCD00010959
CAS Number: 10294-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR