7,8-Dihydroxyflavone hydrate 97%

Supplier: Ambeed
Warning

Synonyms: 7,8-Dihydroxy-2-phenyl-4H-chromen-4-one, 7,8-Dihydroxyflavone

A362609-100MG A362609-25MG A362609-50MG A362609-5G A362609-1G A362609-250MG A362609-25G
76694-220EA 10 USD
76694-220 76694-222 76694-224 76694-226 77572-072 77878-073 77878-074
7,8-Dihydroxyflavone hydrate 97%
7,8-Dihydroxyflavone hydrate

Caution: for Research Use Only

Formula: C₁₅H₁₀O₄
MW: 254.24 g/mol
MDL Number: MFCD00006836
CAS Number: 38183-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR