9-Phenylxanthen-9-ol 98%

Supplier: Ambeed
Warning

Synonyms: Antioxidant agent-15, 9-Phenyl-9H-xanthen-9-ol

A343592-10G A343592-1G A343592-5G
76693-998EA 206.88 USD
76693-998 76694-000 76694-002
9-Phenylxanthen-9-ol 98%
9-Phenylxanthen-9-ol

Caution: for Research Use Only

Formula: C₁₉H₁₄O₂
MW: 274.32 g/mol
MDL Number: MFCD00005058
CAS Number: 596-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR