(R)-2-((6-(3-Aminopiperidin-1-yl)-3-methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)methyl)benzonitrile ≥98%

Supplier: Ambeed
Warning

A295732-10MG A295732-1MG A295732-250MG A295732-25MG A295732-50MG A295732-5G A295732-5MG A295732-100MG A295732-1G
76693-328EA 16 USD
76693-328 76693-330 76693-332 76693-334 76693-336 76693-338 76693-340 77601-324 77601-326
(R)-2-((6-(3-Aminopiperidin-1-yl)-3-methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)methyl)benzonitrile ≥98%
(R)-2-((6-(3-Aminopiperidin-1-yl)-3-methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)methyl)benzonitrile

Caution: for Research Use Only

Formula: C₁₈H₂₁N₅O₂
MDL Number: MFCD09833196
CAS Number: 850649-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR