Cinnamyl acetate 95%

Supplier: Ambeed
Warning

Synonyms: Acetic Acid Cinnamyl Ester

A294685-100G A294685-25G A294685-500G
76693-298EA 13.8 USD
76693-298 76693-300 76693-302
Cinnamyl acetate 95%
Cinnamyl acetate

Caution: for Research Use Only

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
MDL Number: MFCD00008722
CAS Number: 103-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR