Benzohydroxamic acid 98%

Supplier: Ambeed
Warning

A291959-100G A291959-25G A291959-5G
76693-248EA 133 USD
76693-248 76693-250 76693-252
Benzohydroxamic acid 98%
Benzohydroxamic acid

Caution: for Research Use Only

Formula: C₇H₇NO₂
MDL Number: MFCD00002109
CAS Number: 495-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR