β-Carotene 98%

Supplier: Ambeed
Warning

Synonyms: 2,2'-((1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-Tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaene-1,18-diyl)bis(1,3,3-trimethylcyclohex-1-ene), Provitamin A

A286364-100MG A286364-25G A286364-5G A286364-100G A286364-1G A286364-500G
76693-138EA 5.31 USD
76693-138 76693-140 76693-142 77571-780 77626-468 77626-470
β-Carotene 98%
beta-Carotene

Caution: for Research Use Only

Formula: C₄₀H₅₆
MW: 536.89 g/mol
MDL Number: MFCD00001556
CAS Number: 7235-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR