Ethyl-4-oxocyclohexanecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-oxocyclohexanecarboxylate, 4-(Ethoxycarbonyl)-1-cyclohexanone

A257765-100G A257765-10G A257765-1G A257765-25G A257765-500G A257765-5G A257765-1000G
76692-648EA 18 USD
76692-648 76692-650 76692-652 76692-654 76692-656 76692-658 77601-076
Ethyl-4-oxocyclohexanecarboxylate 98%
Ethyl 4-oxocyclohexanecarboxylate

Caution: for Research Use Only

Formula: C₉H₁₄O₃
MW: 170.21 g/mol
MDL Number: MFCD00013285
CAS Number: 17159-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR