Benzoic anhydride 95%

Supplier: Ambeed
Warning

A253041-1000G A253041-100G A253041-25G A253041-500G
76692-548EA 67 USD
76692-548 76692-550 76692-552 76692-554
Benzoic anhydride 95%
Benzoic anhydride

Caution: for Research Use Only

Formula: C₁₄H₁₀O₃
MW: 226.23 g/mol
Boiling Pt: 360
Melting Pt: 40
MDL Number: MFCD00003073
CAS Number: 93-97-0
Merck Index: 13,01093

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR