(3-Bromo-4-methoxyphenyl)acetic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Bromo-4-methoxyphenylacetic acid, 2-(3-Bromo-4-methoxyphenyl)acetic acid
A223232-100G
A223232-1G
A223232-5G
A223232-25G
76691-846EA
130
USD
76691-846
76691-848
76691-850
77626-122
(3-Bromo-4-methoxyphenyl)acetic acid 98%
(3-Bromo-4-methoxyphenyl)acetic acid
Caution: for Research Use Only
Formula:
C₉H₉BrO₃ |
MDL Number:
MFCD00017538 CAS Number: 774-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...