N-Acetyl-D-alanine 97%

Supplier: Ambeed
Warning

A196607-10G A196607-1G A196607-100G A196607-25G A196607-5G
76691-072EA 15 USD
76691-072 76691-074 77600-602 77600-604 77600-606
N-Acetyl-D-alanine 97%
N-Acetyl-D-alanine

Caution: for Research Use Only

Formula: C₅H₉NO₃
MDL Number: MFCD00063133
CAS Number: 19436-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR