4-Amino-L-phenylalanine 95%

Supplier: Ambeed
Warning

A183989-10G A183989-250MG A183989-25G A183989-5G A183989-1G
76690-652EA 33 USD
76690-652 76690-654 76690-656 77600-482 77625-872
4-Amino-L-phenylalanine 95%
4-Amino-L-phenylalanine

Caution: for Research Use Only

Formula: C₉H₁₂N₂O₂
MDL Number: MFCD00069927
CAS Number: 943-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR