7-Methoxycoumarin-4-acetic acid 98%
Supplier: Ambeed
|
Warning
|
A182192-10G
A182192-1G
A182192-250MG
A182192-5G
76690-600EA
134.99
USD
76690-600
76690-602
76690-604
76690-606
7-Methoxycoumarin-4-acetic acid 98%
7-Methoxycoumarin-4-acetic acid
Caution: for Research Use Only
Formula:
C₁₂H₁₀O₅ |
MDL Number:
MFCD00009774 CAS Number: 62935-72-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...