Benzo[d]thiazol-7-amine 97%

Supplier: Ambeed
Warning

A156729-1G A156729-250MG A156729-5G
76689-592EA 66 USD
76689-592 76689-594 76689-596
Benzo[d]thiazol-7-amine 97%
Benzo[d]thiazol-7-amine

Caution: for Research Use Only

Formula: C₇H₆N₂S
MW: 150.2000 g/mol
MDL Number: MFCD10566715
CAS Number: 1123-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR