Butyrophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-Phenylbutan-1-one, Phenyl propyl ketone, 1-Phenyl-1-butanone

A144426-1000G A144426-100G A144426-500G A144426-25G
76689-110EA 167 USD
76689-110 76689-112 76689-114 77299-378
Butyrophenone 98%
Butyrophenone

Caution: for Research Use Only

Formula: C₆H₅C(O)(CH₂)₂CH₃
MDL Number: MFCD00009397
CAS Number: 495-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR