α-Methylcinnamic acid 98%

Supplier: Ambeed
Warning

A108175-100G A108175-10G A108175-25G A108175-500G
76687-330EA 21 USD
76687-330 76687-332 76687-334 76687-336
α-Methylcinnamic acid 98%
α-Methylcinnamic acid

Caution: for Research Use Only

Formula: C₁₀H₁₀O₂
MDL Number: MFCD00002652
CAS Number: 1199-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR