6-TAMRA 98%

Supplier: Ambeed
Warning

Synonyms: 6-Carboxytetramethylrhodamine, 2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-4-carboxybenzoate

A108064-100MG A108064-10MG A108064-1G A108064-1MG A108064-250MG A108064-50MG A108064-5MG A108064-5G
76687-316EA 85 USD
76687-316 76687-318 76687-320 76687-322 76687-324 76687-326 76687-328 77565-902
6-TAMRA 98%
6-TAMRA

Caution: For research use only.

Formula: C₂₅H₂₂N₂O₅
MDL Number: MFCD01073638
CAS Number: 91809-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR