Benzo[b]thiophene ≥98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Thianaphthene, 1-Benzothiophene

OR6572-5G OR6572-25G
76617-208EA 20.87 USD
76617-208 76617-206
Benzo[b]thiophene ≥98%
Benzo[b]thiophene
Formula: C₈H₆S
MW: 134.2 g/mol
Boiling Pt: 221…222 °C
Melting Pt: 28…33 °C
Density: 1.149
Flash Pt: 91 °C
MDL Number: MFCD00005864
CAS Number: 95-15-8
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR