(3-Propylphenyl)boronic acid 97%

Supplier: Ambeed
Warning

A1263388-100MG A1263388-250MG
76680-920EA 36 USD
76680-920 76680-922
(3-Propylphenyl)boronic acid 97%
(3-Propylphenyl)boronic acid

Caution: for Research Use Only

Formula: C₉H₁₃BO₂
MDL Number: MFCD27931626
CAS Number: 891843-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR