Ruthenium(III) acetylacetonate 99% (Ru> 26%)
Supplier: Ambeed
|
Danger
|
Synonyms:
Ruthenium (III) 2,4-pentanedionate, Tris(2,4-pentanedionato-O,O')ruthenium (III), Ruthenium (III) tris(2,4-pentanedionato-O,O'), Tris(2,4-pentanedionato)ruthenium(III)
A1233113-1G
A1233113-250MG
A1233113-25G
A1233113-5G
76679-902EA
23
USD
76679-902
76679-904
76679-906
76679-908
Ruthenium(III) acetylacetonate 99% (Ru> 26%)
Ruthenium (III) acetylacetonate
Caution: for Research Use Only
Formula:
C₁₅H₂₁O₆Ru |
MDL Number:
MFCD00000030 CAS Number: 14284-93-6 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...